The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013098.1 | NP_001013116.1 | 4 - 5 | 739 | 102 |
GenBank mRNA | AB108671.1 | - | 4 - 5 | 739 | 102 |
BC104713.1 | - | 4 - 5 | 645 | 102 |