Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pars2
Entrez Gene ID
313429
Gene Name
prolyl-tRNA synthetase 2, mitochondrial (putative)
Gene Aliases
RGD1305345
Chromosome Location
Chr.5: 126254125 - 126259204 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014064.1 NP_001014086.1 2 - 2 1457 113
XM_006238512.2 XP_006238574.1 2 - 2 1496 113
XM_006238513.2 XP_006238575.1 2 - 2 1307 113
GenBank mRNA BC088403.1 - 2 - 2 1457 113

View Full Product Details