Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lcn11
Entrez Gene ID
100169711
Gene Name
lipocalin 11
Gene Aliases
-
Chromosome Location
Chr.3: 3236843 - 3240131 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135809.1 NP_001129281.1 1 - 2 127 62
GenBank mRNA DQ537495.1 - 1 - 2 127 62

Gene Symbol
LOC100359989
Entrez Gene ID
100359989
Gene Name
lipocalin 11-like
Gene Aliases
-
Chromosome Location
Chr.3: 2935406 - 2938639 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006233654.2 XP_006233716.2 1 - 2 76 62

View Full Product Details