The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012695.3 | NP_036827.3 | 5 - 6 | 779 | 84 |
GenBank mRNA | FQ209484.1 | - | 5 - 6 | 654 | 84 |
FQ209563.1 | - | 5 - 6 | 654 | 84 | |
FQ210641.1 | - | 4 - 5 | 527 | 84 | |
FQ218388.1 | - | 5 - 6 | 655 | 84 | |
FQ219267.1 | - | 5 - 6 | 654 | 84 | |
FQ219345.1 | - | 5 - 6 | 654 | 84 | |
M33329.1 | - | 5 - 6 | 779 | 84 | |
X63410.1 | - | 5 - 6 | 779 | 84 |