Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nckap5l
Entrez Gene ID
315297
Gene Name
NCK-associated protein 5-like
Gene Aliases
RGD1561571
Chromosome Location
Chr.7: 141081943 - 141093924 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271369.1 NP_001258298.1 7 - 8 3236 81

View Full Product Details