Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HIR1
Entrez Gene ID
852275
Gene Name
Gene Aliases
YBL008W
Chromosome Location
Chr.II: 209653 - 212175 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178248.2 NP_009545.2 1 - 1 2452 92

View Full Product Details