Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRX1
Entrez Gene ID
850322
Gene Name
Gene Aliases
YCL035C
Chromosome Location
Chr.III: 60841 - 61173 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178680.1 NP_009895.1 1 - 1 254 76

View Full Product Details