Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPN5
Entrez Gene ID
851408
Gene Name
Gene Aliases
NAS5, YDL147W
Chromosome Location
Chr.IV: 190924 - 192261 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180207.1 NP_010134.1 1 - 1 1150 72

View Full Product Details