Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TFB1
Entrez Gene ID
851906
Gene Name
Gene Aliases
YDR311W
Chromosome Location
Chr.IV: 1085065 - 1086993 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180619.3 NP_010597.3 1 - 1 1843 87

View Full Product Details