Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLX8
Entrez Gene ID
856852
Gene Name
Gene Aliases
YER116C
Chromosome Location
Chr.V: 395348 - 396172 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179006.3 NP_011041.3 1 - 1 648 65

View Full Product Details