Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DAK2
Entrez Gene ID
850491
Gene Name
Gene Aliases
YFL053W
Chromosome Location
Chr.VI: 23423 - 25198 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179914.1 NP_116602.1 1 - 1 1651 65

View Full Product Details