Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPI10
Entrez Gene ID
852735
Gene Name
Gene Aliases
YGL142C
Chromosome Location
Chr.VII: 236269 - 238119 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181007.1 NP_011373.1 1 - 1 1643 90

View Full Product Details