Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAD54
Entrez Gene ID
852713
Gene Name
Gene Aliases
XRS1, YGL163C
Chromosome Location
Chr.VII: 193707 - 196403 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181028.1 NP_011352.1 1 - 1 2247 80

View Full Product Details