Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OKP1
Entrez Gene ID
853090
Gene Name
Gene Aliases
YGR179C
Chromosome Location
Chr.VII: 853675 - 854895 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181308.1 NP_011695.1 1 - 1 1151 86

View Full Product Details