Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEC28
Entrez Gene ID
854734
Gene Name
Gene Aliases
ANU2, YIL076W
Chromosome Location
Chr.IX: 216658 - 217548 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179426.1 NP_012189.2 1 - 1 747 114

View Full Product Details