Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STH1
Entrez Gene ID
854680
Gene Name
Gene Aliases
NPS1, YIL126W
Chromosome Location
Chr.IX: 117992 - 122071 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179474.1 NP_012140.1 1 - 1 3850 90

View Full Product Details