Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IRC24
Entrez Gene ID
854854
Gene Name
Gene Aliases
YIR036C
Chromosome Location
Chr.IX: 422074 - 422865 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179558.3 NP_012302.3 1 - 1 640 71

View Full Product Details