Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHD1
Entrez Gene ID
853823
Gene Name
Gene Aliases
YKL043W
Chromosome Location
Chr.XI: 356748 - 357848 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179609.1 NP_012881.1 1 - 1 851 84

View Full Product Details