Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YNK1
Entrez Gene ID
853798
Gene Name
Gene Aliases
NDK1, YKL067W
Chromosome Location
Chr.XI: 314812 - 315273 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179633.1 NP_012856.1 1 - 1 354 81

View Full Product Details