Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VMA5
Entrez Gene ID
853782
Gene Name
Gene Aliases
CSL5, VAT3, YKL080W
Chromosome Location
Chr.XI: 285030 - 286208 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179646.1 NP_012843.1 1 - 1 952 81

View Full Product Details