Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CWP2
Entrez Gene ID
853765
Gene Name
Gene Aliases
LPR1, YKL096W-A, YKL097W-A
Chromosome Location
Chr.XI: 259253 - 259531 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180025.1 NP_012826.1 1 - 1 152 69

View Full Product Details