Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
REX3
Entrez Gene ID
850797
Gene Name
Gene Aliases
YLR107W
Chromosome Location
Chr.XII: 364116 - 365330 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181994.1 NP_013208.1 1 - 1 1153 89

View Full Product Details