Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EST2
Entrez Gene ID
851028
Gene Name
Gene Aliases
YLR318W
Chromosome Location
Chr.XII: 766542 - 769196 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182207.2 NP_013422.2 1 - 1 2151 95

View Full Product Details