Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DAK1
Entrez Gene ID
854932
Gene Name
Gene Aliases
YML070W
Chromosome Location
Chr.XIII: 133475 - 135229 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182429.1 NP_013641.1 1 - 1 1654 65

View Full Product Details