Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MIH1
Entrez Gene ID
855052
Gene Name
Gene Aliases
YMR036C
Chromosome Location
Chr.XIII: 341856 - 343520 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182533.1 NP_013750.1 1 - 1 1552 73

View Full Product Details