Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TGL3
Entrez Gene ID
855361
Gene Name
Gene Aliases
YMR313C
Chromosome Location
Chr.XIII: 899379 - 901307 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182824.1 NP_014044.1 1 - 1 1853 82

View Full Product Details