Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VID27
Entrez Gene ID
855509
Gene Name
Gene Aliases
YNL212W
Chromosome Location
Chr.XIV: 247461 - 249809 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183050.1 NP_014187.1 1 - 1 2249 102

View Full Product Details