Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AEP3
Entrez Gene ID
856102
Gene Name
Gene Aliases
YPL005W
Chromosome Location
Chr.XVI: 548486 - 550306 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183819.1 NP_015320.1 1 - 1 1752 103

View Full Product Details