Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RGC1
Entrez Gene ID
856231
Gene Name
Gene Aliases
YPR115W
Chromosome Location
Chr.XVI: 754877 - 758128 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184212.1 NP_015440.1 1 - 1 3147 99

View Full Product Details