Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BHLHE40
Entrez Gene ID
100514873
Gene Name
basic helix-loop-helix family member e40
Gene Aliases
-
Chromosome Location
Chr.13: 61526121 - 61533686 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001245010.1 NP_001231939.1 3 - 4 526 70
XM_021068423.1 XP_020924082.1 - 632 70
XM_021068424.1 XP_020924083.1 - 691 70
GenBank mRNA AK345015.1 - 3 - 4 526 70

View Full Product Details