Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYF6
Entrez Gene ID
397005
Gene Name
myogenic factor 6
Gene Aliases
Myf-6
Chromosome Location
Chr.5: 100762918 - 100764771 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244672.1 NP_001231601.1 1 - 2 610 57
GenBank mRNA AY188502.1 - 1 - 2 416 57

View Full Product Details