Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ETFB
Entrez Gene ID
396614
Gene Name
electron transfer flavoprotein subunit beta
Gene Aliases
-
Chromosome Location
Chr.6: 58501892 - 58517580 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205279.1 NP_001192208.1 1 - 2 61 87
GenBank mRNA AK392297.1 - 1 - 2 144 87
AY374470.1 - 1 - 2 61 87

View Full Product Details