Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
UBA1
Entrez Gene ID
100525439
Gene Name
ubiquitin like modifier activating enzyme 1
Gene Aliases
UBE1X
Chromosome Location
Chr.X: 41810726 - 41832818 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013990866.2 XP_013846320.2 - 2151 92
XM_021080354.1 XP_020936013.1 - 2085 92
XM_021080355.1 XP_020936014.1 - 2168 92
XM_021080356.1 XP_020936015.1 - 2123 92
XM_021080357.1 XP_020936016.1 - 1948 92
GenBank mRNA AF132070.1 - 2 - 2 249 92
AK234808.1 - 6 - 6 807 92
AK391881.1 - 16 - 16 2134 92

View Full Product Details