Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ST6GALNAC5
Entrez Gene ID
404690
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 5
Gene Aliases
SIAT7E, ST6GALNAC-V
Chromosome Location
Chr.6: 136207838 - 136391083 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214436.1 NP_999601.1 5 - 5 1150 84
GenBank mRNA AJ620948.2 - 5 - 5 1150 84
AK347308.1 - 5 - 5 1150 84
AK390266.1 - 5 - 5 1180 84

View Full Product Details