Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CREB3
Entrez Gene ID
100142689
Gene Name
cAMP responsive element binding protein 3
Gene Aliases
-
Chromosome Location
Chr.1: 236450573 - 236456381 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123220.1 NP_001116692.1 2 - 3 271 76
XM_005660189.3 XP_005660246.2 - 763 76
XM_005660190.3 XP_005660247.2 - 416 76
XM_013993639.2 XP_013849093.2 - 422 76
GenBank mRNA AK235744.1 - 2 - 3 372 76
AK236157.1 - 2 - 3 388 76
AK389376.1 - 3 - 4 384 76
AK390381.1 - 3 - 4 581 76
AK393011.1 - 2 - 3 400 76
AK400220.1 - 2 - 3 433 76
EU588585.1 - 2 - 3 328 76

View Full Product Details