Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
B2M
Entrez Gene ID
397033
Gene Name
beta-2-microglobulin
Gene Aliases
-
Chromosome Location
Chr.1: 126865767 - 126878501 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213978.1 NP_999143.1 1 - 2 124 60
GenBank mRNA AB436775.1 - 1 - 2 67 60
AK231801.1 - 2 - 3 181 60
AK231841.1 - 1 - 2 95 60
AK344900.1 - 1 - 2 96 60
AK346159.1 - 1 - 2 98 60
AK393822.1 - 1 - 2 98 60
AK394531.1 - - 669 60
AK397591.1 - 1 - 2 124 60
AK398890.1 - 1 - 2 126 60
AK399392.1 - 1 - 2 99 60
AK399739.1 - 1 - 2 97 60
FJ944026.1 - 1 - 2 67 60
L13854.1 - 1 - 2 124 60

Gene Symbol
B2M
Entrez Gene ID
110255236
Gene Name
beta-2-microglobulin
Gene Aliases
-
Chromosome Location
Chr.1: 126837721 - 126844043 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_021096362.1 XP_020952021.1 - 217 60
GenBank mRNA AK239552.1 - 1 - 2 95 60
AK397775.1 - 1 - 2 202 60
AK398958.1 - 1 - 2 94 60
AK399280.1 - 1 - 2 102 60

View Full Product Details