Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFLAR
Entrez Gene ID
414381
Gene Name
CASP8 and FADD like apoptosis regulator
Gene Aliases
C-FLIP, FLIP
Chromosome Location
Chr.15: 104780744 - 104835631 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001628.1 NP_001001628.1 3 - 4 647 76
XM_005652441.3 XP_005652498.1 - 1732 76
XM_005652443.3 XP_005652500.1 - 1561 76
XM_005652444.3 XP_005652501.1 - 1951 76
XM_005652445.3 XP_005652502.1 - 1320 76
XM_005652446.3 XP_005652503.1 - 989 76
XM_005652447.3 XP_005652504.1 - 1149 76
XM_013996333.2 XP_013851787.1 - 2476 76
XM_013996365.2 XP_013851819.1 - 786 76
XM_021074699.1 XP_020930358.1 - 2681 76
GenBank mRNA AB370266.1 - 5 - 6 948 76
AB370267.1 - 5 - 6 948 76
AK234664.1 - 5 - 6 947 76
AK235528.1 - 4 - 5 963 76
AK238641.1 - 3 - 4 447 76
AK389814.1 - 4 - 5 717 76
AK396797.1 - 4 - 5 748 76
AY533020.1 - 3 - 4 647 76
AY533021.1 - 3 - 4 647 76

View Full Product Details