Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MS4A2
Entrez Gene ID
397166
Gene Name
membrane spanning 4-domains A2
Gene Aliases
-
Chromosome Location
Chr.2: 11400415 - 11420314 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214092.2 NP_999257.2 2 - 3 290 86
XM_005660844.3 XP_005660901.2 - 404 86
GenBank mRNA AF525403.1 - 1 - 2 80 86
AK348987.1 - 2 - 3 291 86

View Full Product Details