Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MS4A2
Entrez Gene ID
397166
Gene Name
membrane spanning 4-domains A2
Gene Aliases
-
Chromosome Location
Chr.2: 11400415 - 11420314 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214092.2 NP_999257.2 6 - 7 764 63
GenBank mRNA AF525403.1 - 5 - 6 554 63

View Full Product Details