Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LIN52
Entrez Gene ID
100157325
Gene Name
lin-52 DREAM MuvB core complex component
Gene Aliases
-
Chromosome Location
Chr.7: 97382020 - 97491672 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001185836.1 NP_001172765.1 4 - 5 221 83
XM_005656377.3 XP_005656434.1 - 551 83
XM_005656378.3 XP_005656435.1 - 549 83
GenBank mRNA AK231110.1 - 4 - 5 221 83

View Full Product Details