Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HAO1
Entrez Gene ID
100627803
Gene Name
hydroxyacid oxidase 1
Gene Aliases
-
Chromosome Location
Chr.17: 16746153 - 16802460 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243431.1 NP_001230360.1 2 - 3 333 82
XM_021077275.1 XP_020932934.1 - 329 82
GenBank mRNA AK232269.1 - 2 - 3 317 82
AK393986.1 - 2 - 3 326 82

View Full Product Details