Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYSMD1
Entrez Gene ID
100154705
Gene Name
LysM domain containing 1
Gene Aliases
-
Chromosome Location
Chr.4: 98082551 - 98093427 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243501.1 NP_001230430.1 1 - 2 774 105
GenBank mRNA AK343472.1 - 1 - 2 805 105
EW092615.2 - 1 - 2 452 105

View Full Product Details