Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DHRS7
Entrez Gene ID
100152785
Gene Name
dehydrogenase/reductase 7
Gene Aliases
-
Chromosome Location
Chr.1: 189153579 - 189170745 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244160.1 NP_001231089.1 6 - 7 1310 109
GenBank mRNA AK231851.1 - 6 - 7 1024 109
AK239997.1 - 6 - 7 1064 109
AK398059.1 - 6 - 7 1076 109
AY609687.1 - 6 - 7 1310 109

View Full Product Details