The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244385.1 | NP_001231314.1 | 3 - 4 | 368 | 90 |
GenBank mRNA | AK346675.1 | - | 3 - 4 | 346 | 90 |
AY610472.1 | - | 3 - 4 | 368 | 90 |