Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPID
Entrez Gene ID
100515957
Gene Name
peptidylprolyl isomerase D
Gene Aliases
-
Chromosome Location
Chr.8: 47656269 - 47702805 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244386.1 NP_001231315.1 2 - 3 300 90
XM_021100359.1 XP_020956018.1 - 899 90
GenBank mRNA AK344118.1 - 2 - 3 295 90
AY609657.1 - 2 - 3 300 90

View Full Product Details