Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SBDS
Entrez Gene ID
100516003
Gene Name
SBDS ribosome maturation factor
Gene Aliases
-
Chromosome Location
Chr.3: 16161524 - 16168619 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244393.1 NP_001231322.1 2 - 3 484 102
GenBank mRNA AK238658.1 - 2 - 3 497 102
AK396822.1 - 2 - 3 514 102
AY609615.1 - 2 - 3 484 102

View Full Product Details