Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FADS6
Entrez Gene ID
100513229
Gene Name
fatty acid desaturase 6
Gene Aliases
-
Chromosome Location
Chr.12: 6353716 - 6368987 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244792.1 NP_001231721.1 1 - 2 196 76
GenBank mRNA AK238564.1 - 1 - 2 196 76

View Full Product Details