Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BCH1
Entrez Gene ID
100232875
Gene Name
beta-carotene hydroxylase
Gene Aliases
GSVIVT00002556001
Chromosome Location
Chr.2: 167713 - 169571 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF499108.1 - 2 - 3 435 61

View Full Product Details