Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eif1ax
Entrez Gene ID
448063
Gene Name
eukaryotic translation initiation factor 1A X-linked
Gene Aliases
eif-1a, eif-4c, eif1a, eif1ap1, eif4c, ipp
Chromosome Location
Chr.2: 39393116 - 39401969 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004814.1 NP_001004814.1 3 - 4 436 96
GenBank mRNA BC074588.1 - 3 - 4 436 96
CR760442.2 - 3 - 4 236 96

View Full Product Details