Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
acp1
Entrez Gene ID
448657
Gene Name
acid phosphatase 1
Gene Aliases
acp1-a, acp1-b, haap, lmw-ptp, ptp1a, xlptp1
Chromosome Location
Chr.5: 129694622 - 129703119 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005082.1 NP_001005082.1 4 - 5 320 110
GenBank mRNA BC077004.1 - 4 - 5 320 110

View Full Product Details